2-Fluoro-6-iodobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-6-iodophenyl)methanol

25037-1G 25037-25G 25037-5G
76899-806EA 88.06 USD
76899-806 76899-808 76899-810
2-Fluoro-6-iodobenzyl alcohol ≥97%
2-Fluoro-6-iodobenzyl alcohol
Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09910144
CAS Number: 911825-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR