2-Fluoro-3-methylpyridine-4-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

25022-1G 25022-25G 25022-500MG 25022-5G
76899-792EA 409.51 USD
76899-792 76899-794 76899-796 76899-798
2-Fluoro-3-methylpyridine-4-boronic acid pinacol ester ≥97% research grade
2-Fluoro-3-methylpyridine-4-boronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08063097
CAS Number: 1310383-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR