2-Bromo-4-fluoro-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

25009-1G 25009-500MG 25009-5G
76899-780EA 530.45 USD
76899-780 76899-782 76899-784
2-Bromo-4-fluoro-6-methylphenol ≥97%
2-Bromo-4-fluoro-6-methylphenol
Formula: C₇H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD22414711
CAS Number: 1572185-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR