2-Fluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

24965-25G 24965-5G
76899-772EA 453.01 USD
76899-772 76899-774
2-Fluoro-3-isopropoxybenzoic acid ≥97%
2-Fluoro-3-isopropoxybenzoic acid
Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD28356761
CAS Number: 1346608-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR