2-Methoxy-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

24940-1G 24940-25G 24940-500MG 24940-5G
76899-758EA 264.17 USD
76899-758 76899-760 76899-762 76899-764
2-Methoxy-4-phenylpyridine ≥97%
2-Methoxy-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD11044372
CAS Number: 53698-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR