2-Amino-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Amino-5-methylphenyboronic acid, pinacol ester, 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

24924-1G 24924-25G 24924-500MG 24924-5G
76899-746EA 233.4 USD
76899-746 76899-748 76899-750 76899-752
2-Amino-5-methylphenylboronic acid pinacol ester ≥97%
2-Amino-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795664
CAS Number: 948592-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR