2-Chloro-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

24892-1G 24892-25G 24892-5G
76899-732EA 71.09 USD
76899-732 76899-734 76899-736
2-Chloro-4-phenylpyridine ≥97%
2-Chloro-4-phenylpyridine
Formula: C₁₁H₈ClN
Storage Temperature: Refrigerator
MDL Number: MFCD04114163
CAS Number: 42260-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR