2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Trifluoromethylpyridine-5-boronic acid pinacol ester, 2-(Trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester
24871-1G
24871-25G
24871-5G
76899-702EA
106.09
USD
76899-702
76899-704
76899-706
2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥97%
2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BF₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11226774 CAS Number: 1218790-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...