2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

24851-1G 24851-25G 24851-500MG 24851-5G
76899-676EA 215.37 USD
76899-676 76899-678 76899-680 76899-682
2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester ≥97% research grade
2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12923400
CAS Number: 1083168-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR