2-Methoxy-4-(methylthio)pyridine-3-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24757-1G 24757-500MG 24757-5G
76899-636EA 676.86 USD
76899-636 76899-638 76899-640
2-Methoxy-4-(methylthio)pyridine-3-boronic acid ≥97% research grade
2-Methoxy-4-(methylthio)pyridine-3-boronic acid
Formula: C₇H₁₀BNO₃S
Storage Temperature: Ambient
MDL Number: MFCD22628220
CAS Number: 1451392-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR