2-Fluoro-3-ethoxypyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

24731-1G 24731-500MG 24731-5G
76899-622EA 448.77 USD
76899-622 76899-624 76899-626
2-Fluoro-3-ethoxypyridine-5-boronic acid ≥97%
2-Fluoro-3-ethoxypyridine-5-boronic acid
Formula: C₇H₉BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD26045246
CAS Number: 2121513-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR