2-Fluoro-3-Methylsulfonylpyridine ≥97%

Supplier: AOBChem USA

Warning

24712-1G 24712-25G 24712-5G
76899-612EA 94.43 USD
76899-612 76899-614 76899-616
2-Fluoro-3-Methylsulfonylpyridine ≥97%
2-Fluoro-3-Methylsulfonylpyridine
Formula: C₆H₆FNO₂S
MW: 175.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142584
CAS Number: 1254730-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR