2-Chloro-5-cyclopropylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
24690-1G
24690-25G
24690-5G
76899-598EA
80.63
USD
76899-598
76899-600
76899-602
2-Chloro-5-cyclopropylpyridine ≥97%
2-Chloro-5-cyclopropylpyridine
Precaución: for Research Use Only
Formula:
C₈H₈ClN Storage Temperature: Refrigerator |
MDL Number:
MFCD12963717 CAS Number: 1042986-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...