2-(Benzyloxy)-6-bromonaphthalene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Benzyloxy-6-bromonaphthalene

24655-25G 24655-5G
76899-588EA 381.93 USD
76899-588 76899-590
2-(Benzyloxy)-6-bromonaphthalene ≥97%
2-(Benzyloxy)-6-bromonaphthalene
Formula: C₁₇H₁₃BrO
MW: 313.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01568856
CAS Number: 2234-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR