2-Bromo-6-ethoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

24653-25G 24653-5G
76899-580EA 360.71 USD
76899-580 76899-582
2-Bromo-6-ethoxynaphthalene ≥97%
2-Bromo-6-ethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD01568865
CAS Number: 66217-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR