2-Bromo-4,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24501-1G 24501-25G 24501-500MG 24501-5G
76899-508EA 440.28 USD
76899-508 76899-510 76899-512 76899-514
2-Bromo-4,6-dimethylbenzaldehyde ≥97%
2-Bromo-4,6-dimethylbenzaldehyde
Formula: C₉H₉BrO
MW: 213.071 g/mol
Storage Temperature: Ambient
CAS Number: 88174-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR