2,6-Dibromo-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

24437-1G 24437-25G 24437-500MG 24437-5G
76899-470EA 221.73 USD
76899-470 76899-472 76899-474 76899-476
2,6-Dibromo-4-iodopyridine ≥97%
2,6-Dibromo-4-iodopyridine
Formula: C₅H₂Br₂IN
Storage Temperature: Ambient
MDL Number: MFCD13185521
CAS Number: 1160184-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR