2,6-Bis(1-methylethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

24417-1G 24417-25G 24417-500MG 24417-5G
76899-460EA 260.99 USD
76899-460 76899-462 76899-464 76899-466
2,6-Bis(1-methylethoxy)pyridine ≥97%
2,6-Bis(1-methylethoxy)pyridine
Formula: C₁₁H₁₇NO₂
MW: 195.258 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543699
CAS Number: 1017577-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR