2-Chloro-4-hydroxy-benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-hydroxybenzoic acid methyl ester, Methyl 2-chloro-4-hydroxybenzoate

24318-1G 24318-25G 24318-5G
76899-414EA 66.84 USD
76899-414 76899-416 76899-418
2-Chloro-4-hydroxy-benzoic acid methyl ester ≥97%
2-Chloro-4-hydroxy-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117954
CAS Number: 104253-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR