3-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester, 2-Fluoro-6-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzoic acid methyl ester, Methyl 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Benzoic acid, 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, methyl ester

24273-1G 24273-25G 24273-500MG 24273-5G
76899-398EA 165.51 USD
76899-398 76899-400 76899-402 76899-404
3-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%
3-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996307
CAS Number: 1293284-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR