2-Isopropoxypyrimidine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

24171-1G 24171-25G 24171-500MG 24171-5G
76899-366EA 232.34 USD
76899-366 76899-368 76899-370 76899-372
2-Isopropoxypyrimidine-5-boronic acid pinacol ester ≥97% research grade
2-Isopropoxypyrimidine-5-boronic acid pinacol ester
Formula: C₁₃H₂₁BN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11878341
CAS Number: 1355066-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR