2-Ethoxy-5-pyridineboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxypyridine-5-boronic acid pinacol ester, 2-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

24072-25G 24072-5G
76899-360EA 608.96 USD
76899-360 76899-362
2-Ethoxy-5-pyridineboronic acid pinacol ester ≥95%
2-Ethoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07781182
CAS Number: 1072945-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR