2-Methoxypyrimidin-5-ylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Methoxypyrimidin-5-ylboronic acid pinacol ester

24069-1G 24069-25G 24069-500MG 24069-5G
76899-344EA 149.59 USD
76899-344 76899-346 76899-348 76899-350
2-Methoxypyrimidin-5-ylboronic acid, pinacol ester ≥97%
2-Methoxypyrimidin-5-ylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD13182190
CAS Number: 1052686-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR