2-Fluoro-5-(methoxymethoxy)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

24027-1G 24027-25G 24027-5G
76899-318EA 62.6 USD
76899-318 76899-320 76899-322
2-Fluoro-5-(methoxymethoxy)pyridine ≥97% research grade
2-Fluoro-5-(methoxymethoxy)pyridine
Formula: C₇H₈FNO₂
MW: 157.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123917
CAS Number: 1034467-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR