2-Fluoro-3-(methylthio)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

24012-25G 24012-5G
76899-306EA 524.09 USD
76899-306 76899-308
2-Fluoro-3-(methylthio)pyridine ≥97% research grade
2-Fluoro-3-(methylthio)pyridine
Formula: C₆H₆FNS
MW: 143.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543684
CAS Number: 210992-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR