2-Bromo-3-methyl-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

24005-25G 24005-5G
76899-296EA 524.09 USD
76899-296 76899-298
2-Bromo-3-methyl-5-(methylthio)pyridine ≥97%
2-Bromo-3-methyl-5-(methylthio)pyridine
Formula: C₇H₈BrNS
MW: 218.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403705
CAS Number: 2624417-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR