2,5-Dibromo-4-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

23984-1G 23984-25G 23984-500MG 23984-5G
76899-284EA 157.02 USD
76899-284 76899-286 76899-288 76899-290
2,5-Dibromo-4-methoxypyridine ≥97% research grade
2,5-Dibromo-4-methoxypyridine
Formula: C₆H₅Br₂NO
MW: 266.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18255776
CAS Number: 1211520-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR