(6-Phenoxypyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

23981-1G 23981-25G 23981-500MG 23981-5G
76899-268EA 239.77 USD
76899-268 76899-270 76899-272 76899-274
(6-Phenoxypyridin-3-yl)boronic acid ≥97%
(6-Phenoxypyridin-3-yl)boronic acid
Formula: C₁₁H₁₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD12403506
CAS Number: 1270921-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR