2,5-Dimethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

23890-1G 23890-25G 23890-5G
76899-228EA 64.72 USD
76899-228 76899-230 76899-232
2,5-Dimethoxyphenol ≥97%
2,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00094237
CAS Number: 18113-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR