2'-Bromo-2,6-dimethoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

23821-25G 23821-5G
76899-218EA 204.76 USD
76899-218 76899-220
2'-Bromo-2,6-dimethoxy-1,1'-biphenyl ≥95%
2'-Bromo-2,6-dimethoxy-1,1'-biphenyl
Formula: C₁₄H₁₃BrO₂
MW: 293.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369791
CAS Number: 755017-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR