2-Methoxy-4-(methylthio)pyridine-5-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23755-1G 23755-25G 23755-500MG 23755-5G
76899-202EA 439.22 USD
76899-202 76899-204 76899-206 76899-208
2-Methoxy-4-(methylthio)pyridine-5-boronic acid ≥97% research grade
2-Methoxy-4-(methylthio)pyridine-5-boronic acid
Formula: C₇H₁₀BNO₃S
Storage Temperature: Ambient
MDL Number: MFCD22543669
CAS Number: 1451392-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR