2,5-Dichloro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

23739-1G 23739-25G 23739-500MG 23739-5G
76899-186EA 203.7 USD
76899-186 76899-188 76899-190 76899-192
2,5-Dichloro-4-(methylthio)pyridine ≥97%
2,5-Dichloro-4-(methylthio)pyridine
Formula: C₆H₅Cl₂NS
MW: 194.082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32706997
CAS Number: 474824-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR