2-Bromo-6-fluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-fluorophenyl methyl sulphide, 1-Bromo-3-fluoro-2-(methylsulphanyl)benzene

23655-1G 23655-25G 23655-5G
76899-138EA 70.02 USD
76899-138 76899-140 76899-142
2-Bromo-6-fluorothioanisole ≥97%
2-Bromo-6-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFS
Storage Temperature: Refrigerator
MDL Number: MFCD22056775
CAS Number: 1370025-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR