2-Isopropoxy-4-methylpyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

23405-1G 23405-500MG 23405-5G 23405-25G
76899-042EA 462.56 USD
76899-042 76899-044 76899-046 77654-302
2-Isopropoxy-4-methylpyridine-5-boronic acid pinacol ester ≥97% research grade
2-Isopropoxy-4-methylpyridine-5-boronic acid pinacol ester
Formula: C₁₅H₂₄BNO₃
Storage Temperature: Ambient
MDL Number: MFCD16996674
CAS Number: 1451391-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR