2-Chloro-5-methoxy-3-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
23081-1G
23081-25G
23081-500MG
23081-5G
76899-002EA
251.44
USD
76899-002
76899-004
76899-006
76899-008
2-Chloro-5-methoxy-3-methylpyridine ≥97%
2-Chloro-5-methoxy-3-methylpyridine
Formula:
C₇H₈ClNO Storage Temperature: Refrigerator |
MDL Number:
MFCD16610592 CAS Number: 74650-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...