2,6-Dimethyl-4-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
23026-25G
23026-5G
76898-978EA
275.84
USD
76898-978
76898-980
2,6-Dimethyl-4-methoxybenzoic acid ≥97%
2,6-Dimethyl-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ MW: 180.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01194287 CAS Number: 37934-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...