2-(Methylthio)benzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

23009-1G 23009-25G 23009-5G
76898-964EA 79.57 USD
76898-964 76898-966 76898-968
2-(Methylthio)benzotrifluoride ≥95%
2-(Methylthio)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₇F₃S
Storage Temperature: Refrigerator
MDL Number: MFCD01882714
CAS Number: 322-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR