(4-(Benzyloxy)-2,6-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethyl-4-(phenylmethoxy)phenylboronic acid

22665-1G 22665-25G 22665-500MG 22665-5G
76898-872EA 165.51 USD
76898-872 76898-874 76898-876 76898-878
(4-(Benzyloxy)-2,6-dimethylphenyl)boronic acid ≥97%
(4-(Benzyloxy)-2,6-dimethylphenyl)boronic acid
Formula: C₁₅H₁₇BO₃
MW: 256.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194673
CAS Number: 865139-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR