2-Bromo-5-ethoxy-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

22661-1G 22661-25G 22661-5G
76898-858EA 62.6 USD
76898-858 76898-860 76898-862
2-Bromo-5-ethoxy-1,3-dimethylbenzene ≥95%
2-Bromo-5-ethoxy-1,3-dimethylbenzene
Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD18917789
CAS Number: 91799-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR