2-Fluoro-3-methoxyphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-methoxyphenylboronic acid pinacol ester, 2-(2-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

22600-25G 22600-5G
76898-824EA 310.85 USD
76898-824 76898-826
2-Fluoro-3-methoxyphenylboronic acid, pinacol ester ≥97%
2-Fluoro-3-methoxyphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618962
CAS Number: 1165936-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR