6-Chloro-4-iodo-3-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

22596-1G 22596-25G 22596-500MG 22596-5G
76898-808EA 168.69 USD
76898-808 76898-810 76898-812 76898-814
6-Chloro-4-iodo-3-hydroxypyridine ≥97%
6-Chloro-4-iodo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃ClINO
Storage Temperature: Refrigerator
MDL Number: MFCD11977445
CAS Number: 877133-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR