2-Fluoro-3-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

22585-25G 22585-5G
76898-790EA 346.92 USD
76898-790 76898-792
2-Fluoro-3-iodobenzoic acid ≥95%
2-Fluoro-3-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
Storage Temperature: Ambient
MDL Number: MFCD07774248
CAS Number: 447464-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR