2,5-Dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

22534-25G 22534-5G
76898-732EA 272.66 USD
76898-732 76898-734
2,5-Dimethoxypyridine ≥97%
2,5-Dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12546620
CAS Number: 867267-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR