2-Bromo-6-propoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

22499-25G 22499-5G
76898-726EA 524.09 USD
76898-726 76898-728
2-Bromo-6-propoxynaphthalene ≥97%
2-Bromo-6-propoxynaphthalene
Formula: C₁₃H₁₃BrO
MW: 265.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18262543
CAS Number: 97476-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR