(2,5-Dichloro-3-pyridinyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichloropyridine-3-methanol

22480-1G 22480-25G 22480-5G
76898-714EA 81.69 USD
76898-714 76898-716 76898-718
(2,5-Dichloro-3-pyridinyl)methanol ≥97%
(2,5-Dichloro-3-pyridinyl)methanol
Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659487
CAS Number: 558465-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR