3-((4-methylpiperazin-1-yl)methyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22404-1G 22404-25G 22404-500MG 22404-5G
76898-660EA 222.79 USD
76898-660 76898-662 76898-664 76898-666
3-((4-methylpiperazin-1-yl)methyl)phenylboronic acid ≥97%
3-((4-methylpiperazin-1-yl)methyl)phenylboronic acid
Formula: C₁₂H₁₉BN₂O₂
MW: 234.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16198076
CAS Number: 1171044-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR