2-Acetylthiophene-3-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Acetyl-3-thiopheneboronic acid, 2-Acetyl-3-thienylboronic acid, (2-Acetylthiophen-3-yl)boronic acid

22350-25G 22350-5G
76898-644EA 488.02 USD
76898-644 76898-646
2-Acetylthiophene-3-boronic acid ≥97% research grade
2-Acetylthiophene-3-boronic acid
Formula: C₆H₇BO₃S
Storage Temperature: Ambient
MDL Number: MFCD01318932
CAS Number: 36155-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR