2-Fluoro-4-methylanisole ≥97%

Supplier: AOBChem USA

Warning

22075-25G 22075-5G
76898-584EA 194.15 USD
76898-584 76898-586
2-Fluoro-4-methylanisole ≥97%
2-Fluoro-4-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060270
CAS Number: 399-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR