2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Bis(trifluoromethyl)-6-methoxybenzaldehyde, 2-Methoxy-4,6-bis(trifluoromethyl)benzaldehyde

22060-25G 22060-5G
76898-578EA 630.18 USD
76898-578 76898-580
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde ≥95%
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆F₆O₂
MW: 272.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660316
CAS Number: 180134-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR