2-Bromo-6-(methoxymethoxy)naphthalene ≥97% research grade

Supplier: AOBChem USA

Warning

22019-25G 22019-5G
76898-568EA 304.48 USD
76898-568 76898-570
2-Bromo-6-(methoxymethoxy)naphthalene ≥97% research grade
2-Bromo-6-(methoxymethoxy)naphthalene
Formula: C₁₂H₁₁BrO₂
Storage Temperature: Refrigerator
CAS Number: 111359-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR